
Isobutyric acid, Supelco®
Catalog # 46935-U
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- Seq:2725
- Pk Info:500 mg
Specifications
About this item
:
:88.11 g/mol
Merck Index:13,05172
Boiling Pt:155 °C (1013 hPa)
Melting Pt:–45 °C
Density:0.949 g/cm³ (20 °C)
Flash Pt:57 °C
Storage Temperature:Ambient
EINECS:201-195-7
UN:2529
ADR:3,III
:M-Clarity™ quality level = MQ100
:GHS07
CAS Number:79-31-2
Formula:C₄H₈O₂
MDL Number:MFCD00002658