Order Entry
Denmark
ContactUsLinkComponent
Isobutyric acid
Irritant
Isobutyric acid
Catalog # EHERC14395500
Supplier:  EHRENSTORFER
CAS Number:  
Irritant
Isobutyric acid
Catalog # EHERC14395500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :88.11 g/mol
Merck Index:13,05172
Boiling Pt:155 °C (1013 hPa)
Melting Pt:–45 °C
Density:0.949 g/cm³ (20 °C)
Flash Pt:57 °C
Storage Temperature:Ambient
EINECS:201-195-7
UN:2529
ADR:3,III
 :GHS07
CAS Number:79-31-2
Formula:C₄H₈O₂
MDL Number:MFCD00002658