Order Entry
Denmark
Orders LinkContactUsLinkComponent
2-Mercaptoethanol

2-Mercaptoethanol

Toxic
Corrosive
Environment
Marketsource
Supplier:   Roth Carl
2-Mercaptoethanol
Supplier:  Roth Carl
Marketsource
Certificates
2-Mercaptoethanol
2-Mercaptoethanol

2 Options Available Below

Product Details & Documents
Synonyms:  2-Hydroxyethyl mercaptan, β-Mercaptoethanol
  • CAS Number:60-24-2
  • Formula:HSCH₂CH₂OH
  • MDL Number:MFCD00004890
  • Molecular Weight:78.14 g/mol
  • Merck Index:13,05894
  • Boiling Pt:157 °C (1013 hPa)
  • Melting Pt:–100 °C
  • Density:1.12 g/cm³ (20 °C)
  • Flash Pt:68 °C
  • Storage Temperature:Ambient
  • EINECS:200-464-6
  • UN:2966
  • ADR:6.1,II
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.