Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

2-Mercaptoethanol

Glass bottle, 100 mL
Catalog # A1108.0100
Toxic
Corrosive
Environment
Supplier:   PanReac AppliChem
2-Mercaptoethanol
Catalog # A1108.0100
Supplier:  PanReac AppliChem
CAS Number:  60-24-2
2-Mercaptoethanol
2-Mercaptoethanol

Glass bottle, 100 mL

CAS Number:  60-24-2
Price
485,00 kr
Log in for availability
1 * 100 mL
Product Details & Documents
Synonyms:  2-Hydroxyethyl mercaptan, β-Mercaptoethanol
  • CAS Number:60-24-2
  • Formula:HSCH₂CH₂OH
  • MDL Number:MFCD00004890
  • Molecular Weight:78.14 g/mol
  • Merck Index:13,05894
  • Boiling Pt:157 °C (1013 hPa)
  • Melting Pt:–100 °C
  • Density:1.12 g/cm³ (20 °C)
  • Flash Pt:68 °C
  • Storage Temperature:Ambient
  • EINECS:200-464-6
  • UN:2966
  • ADR:6.1,II

 

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.