Methanol ≥99.9%, CHROMASOLV™ for GC/MS analysis of volatile organics, Riedel-de Haën™
Catalog # 414816-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Specifications
Specification Test Results
- Autoignition Temperature:725 °F
- IR spectrum:complying
- Assay (GC):Min. 99.90 %
- Non-volatile matter:Max. 1 ppm
- Water (Karl Fischer):Max. 0.2 %
- Impurity Profile:complying
- Vapor Pressure:410 mmHg ( 50 °C)
- Refractive Index:n20/D 1.329 (lit.)
Specification Test Results
About this item
Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
:
:32.04 g/mol
Merck Index:13,05984
Boiling Pt:64…65 °C (1.013 hPa)
Melting Pt:–98 °C
Density:0.79 g/cm³ (20 °C)
Flash Pt:11 °C
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
REACH:01-2119433307-44
:GHS08
:GHS02
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595