Order Entry
Denmark
ContactUsLinkComponent
 
 
Propionic anhydride 99%
 
undefined
 
Propionic anhydride 99%
 

 

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4030 to 1.4050 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Color scale:
    ≤20 APHA

 

 

Frequently Bought Together