


Acetic anhydride, anhydrous 99+%, pure
:108-24-7
:
:(CH₃CO)₂O
:MFCD00008705
:13,00057
:139.9 °C (1013 hPa)
:–73.1 °C
:1.087 g/cm³
:54 °C
:1715
:8,II
:102.09 g/mol
:Ambient
:203-564-8
:01-2119486470-36
:GHS02
:GHS05
:GHS06
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3890 to 1.3910 (20°C, 589 nm)
- GC:≥99.0 %
- Specific optical rotation:+24° to +28° (20°C, 589 nm) (c=1 in CHCl3)
Frequently Bought Together