1,6-Diisocyanatohexane ≥98%
: Hexamethylene diisocyanate
:822-06-0
:
:C₈H₁₂N₂O₂
:MFCD00002047
:255 °C
:–67 °C
:1.05
:168.2 g/mol
:130 °C (266 °F)
:Ambient
:2281
:6.1 II
:Packaged under argon.
:212-485-8
:GHS06
:GHS08
:GHS07
- Environmentally Preferable:
Frequently Bought Together