N,N,N',N'-Tetramethylethylenediamine (TEMED)
Catalog # SHETIB70120
Supplier: IBI SCIENTIFIC
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:116.21 g/mol
Boiling Pt:120 °C (1013 hPa)
Melting Pt:–64 °C
Density:0.7765 g/cm³ (20 °C)
Flash Pt:17 °C (closed cup)
Storage Temperature:Ambient
EINECS:203-744-6
UN:2372
ADR:3,II
:GHS02
:GHS05
:GHS07
CAS Number:110-18-9
Formula:C₆H₁₆N₂
MDL Number:MFCD00008335