Order Entry
Denmark
ContactUsLinkComponent
N,N,N',N'-Tetramethylethylenediamine (TEMED) 99%, extra pure
N,N,N',N'-Tetramethylethylenediamine (TEMED) 99%, extra pure
Certificates
undefined
N,N,N',N'-Tetramethylethylenediamine (TEMED) 99%, extra pure

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4169 to 1.4189 (20°C, 589 nm)
  • Water:
    ≤1 % (K.F.)
  • GC:
    ≥98.5 %
  • Color scale:
    ≤100 APHA

Specification Test Results

Related Information

Frequently Bought Together