Triethanolamine (trolamine) 99+%
:102-71-6
:
:C₆H₁₅NO₃
:MFCD00002855
:149.19 g/mol
:13,09739
:360 °C (760 mmHg)
:21 °C
:1.1245 g/cm³
:190 °C
:Ambient
:203-049-8
- Appearance:Clear colorless to light yellow Viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.4830 to 1.4860 (20°C, 589 nm)
- Titration with HClO4:≥99.0 %
Frequently Bought Together