

Pentafluoropropionic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:422-64-0
:
Formula:C₃HF₅O₂
MDL Number:MFCD00004170
:164.03 g/mol
Boiling Pt:96 °C (1013 hPa)
Density:1.561 g/cm³ (20 °C)
EINECS:207-021-6
UN:2922
ADR:8 (6.1),II
:M-Clarity™ quality level = MQ200
:GHS06
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) soluble
- Molar Mass:164.03 g/mol
- Boiling Point:96 - 97 °C (984 hPa)
- Vapor Pressure:52 hPa (25 °C)
- Refractive Index:1.2838 (25 °C, 589 nm)
- Density:1.57 g/cm3 (20 °C)
Specification Test Results
Related Information
Frequently Bought Together