Thiourea
Marketsource
Supplier: Roth Carl
Jump to:
About this item
Synonyms: Thiocarbamide, Sulphourea
- CAS Number:62-56-6
- Formula:NH₂CSNH₂
- MDL Number:MFCD00008067
- Molecular Weight:76.12 g/mol
- Merck Index:13,09443
- Melting Pt:174…177 °C
- Density:1.435 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-543-5
- UN:3077
- ADR:9,III
2 Options Available Below
Product Details & Documents
Synonyms: Thiocarbamide, Sulphourea
- CAS Number:62-56-6
- Formula:NH₂CSNH₂
- MDL Number:MFCD00008067
- Molecular Weight:76.12 g/mol
- Merck Index:13,09443
- Melting Pt:174…177 °C
- Density:1.435 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-543-5
- UN:3077
- ADR:9,III
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource



