Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Thiourea ≥99%

Health
Irritant
Environment
Thiourea ≥99%
Certificates
Thiourea ≥99%

4 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 62-56-6 Molecular Formula: CH4N2S Formula Weight: 76.12 Odour: Odourless Melting Point: 172-178° Beilstein Reference: 605327 Merck Reference: 14,9367 UN No.: 3077 MDL No.: MFCD00008067

Synonyms:  Thiocarbamide, Sulphourea
  • CAS Number:62-56-6
  • Formula:NH₂CSNH₂
  • MDL Number:MFCD00008067
  • Molecular Weight:76.12 g/mol
  • Merck Index:14,9367
  • Melting Pt:172…178 °C
  • Density:1.405
  • Storage Temperature:Ambient
  • EINECS:200-543-5
  • UN:3077
  • ADR:9,III
Product Family Options

Product Information

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.