

EDTA tetrasodium salt tetrahydrate 99.5% for molecular biology DNase-, RNase-, protease-free
Catalog # ACRO327211000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Titration Complexometric:≥99.4 %
- Solubility:(0.1 M in water) Clear colorless
- UV:(0.1 M in water)
- pH:10.7 to 11.7 (0.1 M in water)
- Dnase,Rnase,Protease activity:Not detected
- Iron (Fe):≤50 ppm
- Heavy metals (as Pb):≤10 ppm
Specification Test Results
About this item
:
:452.23 g/mol
Merck Index:12,03557
Melting Pt:> 300 °C
Density:6,9 g/cm³ (20 °C)
Flash Pt:>100 °C
EINECS:200-573-9
:GHS07
:GHS05
Storage Temperature:Ambient
CAS Number:13235-36-4
Formula:C₁₀H₁₂N₂O₈Na₄·4H₂O
MDL Number:MFCD00150027