
EDTA (ethylenediamine tetraacetic acid) ≥99.0% for complexometry, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:60-00-4
:
Formula:C₁₀H₁₆N₂O₈
MDL Number:MFCD00003541
:292.25 g/mol
Merck Index:13,03546
Boiling Pt:614 °C (1013 hPa)
Melting Pt:240 °C
Density:0.86 g/cm³ (20 °C)
Flash Pt:≥ 100 °C
Storage Temperature:Ambient
EINECS:200-449-4
REACH:01-2119486399-18
:GHS07
Specification Test Results
- Assay:99.0 - 101.0 %
- Identity (IR):complying
- Residue on ignition:Max. 0.1 %
- Colour of the solution:complying
- Clearness of the solution:complying
- Calcium (Ca):Max. 10 ppm
- Cadmium (Cd):Max. 5 ppm
- Cobalt (Co):Max. 5 ppm
- Chromium (Cr):Max. 5 ppm
- Copper (Cu):Max. 5 ppm
- Iron (Fe):Max. 5 ppm
- Potassium (K):Max. 50 ppm
- Magnesium (Mg):Max. 5 ppm
- Manganese (Mn):Max. 5 ppm
- Sodium (Na):Max. 500 ppm
- Nickel (Ni):Max. 5 ppm
- Lead (Pb):Max. 5 ppm
- Zinc (Zn):Max. 5 ppm
- Chloride (Cl):Max. 50 ppm
Specification Test Results
Frequently Bought Together