
Diammonium hydrogen citrate
Supplier: Roth Carl
Certificates
About this item
CAS Number:3012-65-5
:
Formula:HOC(CO₂H)(CH₂CO₂NH₄)₂
MDL Number:MFCD00013068
:226.19 g/mol
Merck Index:13,00513
Density:1.483 g/cm³ (20 °C)
EINECS:221-146-3
:GHS07
Storage Temperature:Ambient
Frequently Bought Together