
Diammonium hydrogen citrate, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 0020.1000
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Assay (by formol method):98.0 - 103.0 %
- Chloride (Cl):max. 0.001 %
- Iron (Fe):max. 0.001 %
- Heavy Metals (as Pb):max. 0.0005 %
- Insoluble Matter:max. 0.005 %
- Oxalate (C2O4):passes test
- Phosphate (PO4):max. 0.0005 %
- Residue after Ignition:max. 0.01 %
- Sulfur Compounds (as SO4):max. 0.005 %
Specification Test Results
About this item
:
:226.19 g/mol
Merck Index:13,00513
Density:1.483 g/cm³ (20 °C)
EINECS:221-146-3
:GHS07
Storage Temperature:Ambient
CAS Number:3012-65-5
Formula:HOC(CO₂H)(CH₂CO₂NH₄)₂
MDL Number:MFCD00013068