Order Entry
Denmark
Orders LinkContactUsLinkComponent
Glycine
Marketsource
 :   Roth Carl
Glycine
 :  Roth Carl
Marketsource
Glycine
Glycine
 :  Aminoacetic acid, Glycocoll, H-Gly-OH
  •  :56-40-6
  •  :NH₂CH₂COOH
  •  :MFCD00008131
  •  :75.07 g/mol
  •  :13,04504
  •  :181 °C (1013 hPa)
  •  :83 °C
  •  :1.64 g/cm³ (20 °C)
  •  :Ambient
  •  :200-272-2
Product Family Options
  • Pack type
    Pk
  • Glass bottle for solids
    250 g
    1 * 250 g
    1 * 250 g - 235,23 kr
    235,23 kr
    1 * 250 g
       235,23 kr
    Marketsource
    Marketsource
  • Glass bottle for solids
    1 kg
    1 * 1 kg
    1 * 1 kg - 746,02 kr
    746,02 kr
    1 * 1 kg
       746,02 kr
    Marketsource
    Marketsource
Anbefalinger