Order Entry
Denmark
Orders LinkContactUsLinkComponent
Glycine

Glycine

Glass bottle for solids, 1 kg
Catalog # RESERES9582G-A103X
Supplier:   RESEARCH ORGANICS
Glycine
Catalog # RESERES9582G-A103X
Supplier:  RESEARCH ORGANICS
CAS Number:  56-40-6
Glycine
Glycine

Glass bottle for solids, 1 kg

CAS Number:  56-40-6
Price
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Aminoacetic acid, Glycocoll, H-Gly-OH
  • CAS Number:56-40-6
  • Formula:NH₂CH₂COOH
  • MDL Number:MFCD00008131
  • Molecular Weight:75.07 g/mol
  • Merck Index:13,04504
  • Boiling Pt:181 °C (1013 hPa)
  • Melting Pt:83 °C
  • Density:1.64 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-272-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications