Glycine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH
CAS Number:56-40-6
:
Formula:NH₂CH₂COOH
MDL Number:MFCD00008131
Boiling Pt:181
Melting Pt:83
Density:1.64 g/cm³ (20 °C)
Molecular Weight:75.07 g/mol
Merck Index:13,04504
Storage Temperature:Ambient
EINECS:200-272-2
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:Min. 98 %
- Ignition residue (SO4):Max. 0.2 %
- Cl (Chloride):Max. 0.02 %
- SO4 (Sulphate):Max. 0.02 %
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000)
Packaging Information
Frequently Bought Together