Glycine, BAKER ANALYZED® for biotechnology, J.T.Baker®
: Avantor
: Aminoacetic acid, Glycocoll, H-Gly-OH
:56-40-6
:
:NH₂CH₂COOH
:MFCD00008131
:181
:83
:1.64 g/cm³ (20 °C)
:75.07 g/mol
:13,04504
:Ambient
:200-272-2
- Environmentally Preferable:
- Assay (by Perchloric Acid titrn.):min. 99 %
- Heavy Metals (as Pb):max. 0.002 %
- Homogenity (by TLC):passes test
- Loss on Drying at 105°C:max. 0.2 %
- Readily Carbonisable Substances:passes test
- Residue after Ignition:max. 0.1 %
- Trace Impurities (in ppm) Arsenic (As):max. 3
- Trace Impurities (in ppm) Iron (Fe):max. 5
Frequently Bought Together