Glycine ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99.5+% CAS No.: 56-40-6 Molecular Formula: H2NCH2CO2H Formula Weight: 75.07 Physical Form: Powder Density: 1.595 pH: 5,97 Melting Point: ca 245° dec. Beilstein Reference: 635782 Merck Reference: 14,4491 MDL No.: MFCD00008131
Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH
- CAS Number:56-40-6
- Formula:NH₂CH₂COOH
- MDL Number:MFCD00008131
- Molecular Weight:75.07 g/mol
- Merck Index:14,4491
- Boiling Pt:181 °C (1013 hPa)
- Melting Pt:∼ 245 °C (dec.)
- Density:1.595
- Storage Temperature:Ambient
- EINECS:200-272-2
2 Options Available Below
Product Details & Documents
Purity: 99.5+% CAS No.: 56-40-6 Molecular Formula: H2NCH2CO2H Formula Weight: 75.07 Physical Form: Powder Density: 1.595 pH: 5,97 Melting Point: ca 245° dec. Beilstein Reference: 635782 Merck Reference: 14,4491 MDL No.: MFCD00008131
Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH
- CAS Number:56-40-6
- Formula:NH₂CH₂COOH
- MDL Number:MFCD00008131
- Molecular Weight:75.07 g/mol
- Merck Index:14,4491
- Boiling Pt:181 °C (1013 hPa)
- Melting Pt:∼ 245 °C (dec.)
- Density:1.595
- Storage Temperature:Ambient
- EINECS:200-272-2
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



