Order Entry
Denmark
ContactUsLinkComponent
 
Glycine 98%, extra pure
 
undefined
Glycine 98%, extra pure
 

 

  • Environmentally Preferable:

 

 

  • Appearance:
    White to off-white Powder or crystals
  • Infrared spectrum:
    Conforms
  • pH:
    5.5 to 6.5 (5 % in water)
  • Titration with HClO4:
    ≥97.5 %

 

 

Frequently Bought Together