Order Entry
Denmark
ContactUsLinkComponent
Ammonium formate ≥99% (by anhydrous basis) eluent additive, for LC-MS, Fluka™
Irritant
Ammonium formate ≥99% (by anhydrous basis) eluent additive, for LC-MS, Fluka™
Catalog # 55674-50G
CAS Number:  
Irritant
Ammonium formate ≥99% (by anhydrous basis) eluent additive, for LC-MS, Fluka™
Catalog # 55674-50G
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Assay (calculated on the anhydrous basis):
    99.0 - 101.0 %
  • Water (Karl Fischer):
    Max. 2 %
  • Sulfated ash:
    Max. 0.1 %
  • Aluminium (Al):
    Max. 1 ppm
  • Arsenic (As):
    Max. 0.1 ppm
  • Barium (Ba):
    Max. 1 ppm
  • Bismuth (Bi):
    Max. 5 ppm
  • Calcium (Ca):
    Max. 5 ppm
  • Cadmium (Cd):
    Max. 1 ppm
  • Cobalt (Co):
    Max. 1 ppm
  • Chromium (Cr):
    Max. 1 ppm
  • Copper (Cu):
    Max. 1 ppm
  • Iron (Fe):
    Max. 1 ppm
  • Potassium (K):
    Max. 5 ppm
  • Lithium (Li):
    Max. 5 ppm
  • Magnesium (Mg):
    Max. 1 ppm
  • Manganese (Mn):
    Max. 1 ppm
  • Molybdenum (Mo):
    Max. 5 ppm
  • Sodium (Na):
    Max. 5 ppm
  • Nickel (Ni):
    Max. 5 ppm
  • Lead (Pb):
    Max. 5 ppm
  • Strontium (Sr):
    Max. 1 ppm
  • Zinc (Zn):
    Max. 1 ppm
  • Chloride (Cl):
    Max. 50 ppm
  • Total S (as SO₄):
    Max. 50 ppm
  • Suitability for the LC-MS:
    complying
  • Appearance of the solution:
    complying
  • Solution 1 M in H₂O:
    complying

Specification Test Results

About this item

 :
 :63.06 g/mol
Merck Index:13,00525
Melting Pt:116 °C
Density:1.266 g/cm³ (20 °C)
EINECS:208-753-9
 :GHS07
Storage Temperature:Ambient
CAS Number:540-69-2
Formula:HCO₂NH₄
MDL Number:MFCD00013103