Parteck® LUB CST (Calcium stearate), EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF, FCC vegetable grade, SAFC®
Catalog # 1.00664.9020
Supplier: MERCK PRODUCTION CHEMICALS
CAS Number:
Specifications
- Pack type:Cardboard carton
- Pk:20 kg
- Seq:4241
Specifications
About this item
:
:607.03 g/mol
Merck Index:13,01708
Melting Pt:147…149 °C
Density:1.065 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:216-472-8
CAS Number:1592-23-0
:66071-81-6
Formula:C₃₆H₇₀CaO₄
MDL Number:MFCD00036390