Order Entry
Denmark
Orders LinkContactUsLinkComponent
Sodium acetate

Sodium acetate

Marketsource
Supplier:   Roth Carl
Sodium acetate
Supplier:  Roth Carl
Marketsource
Certificates

4 Options Available Below

Product Details & Documents
Synonyms:  Acetic acid sodium salt
  • CAS Number:127-09-3
  • Formula:H₃CCOONa
  • MDL Number:MFCD00012459
  • Molecular Weight:82.03 g/mol
  • Merck Index:12,08711
  • Boiling Pt:881.4 °C (1013 hPa)
  • Melting Pt:324 °C
  • Density:1.528 g/cm³ (20 °C)
  • Flash Pt:>250 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:204-823-8
  • REACH:01-2119485123-42
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg

    Specifications:
    Marketsource
    Marketsource
  • 25 kg
    Log in for availability
    Log in for availability
    1 * 25 kg
    1 * 25 kg

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.