Valeric acid
Glass bottle, 250 mL
Catalog # ROTH4874.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Pentanoic acid
- CAS Number:109-52-4
- Formula:C₅H₁₀O₂
- MDL Number:MFCD00004413
- Molecular Weight:102.13 g/mol
- Merck Index:12,10042
- Boiling Pt:186 °C (1013 hPa)
- Melting Pt:–32 °C
- Density:1.11 g/cm³ (20 °C)
- Flash Pt:89 °C
- Storage Temperature:Ambient
- EINECS:203-677-2
- UN:3265
- ADR:8,III
Specifications
Specifications
- Pack type:Glass bottle
- Pk:250 mL



