Dibutyl sebacate
Glass bottle for solids, 500 mg
Catalog # PROOMM0925.00
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
Synonyms: Sebacic acid dibutyl ester, 1,10-Decanedioic acid dibutyl ester, Dibutyl decanedioate
- CAS Number:109-43-3
- Formula:C₁₈H₃₄O₄
- MDL Number:MFCD00027218
- Molecular Weight:314.47 g/mol
- Boiling Pt:180 °C (1013 hPa)
- Melting Pt:–11 °C
- Density:0,94 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:203-672-5
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



