Supelco® Organic Reference Standard, Methyl stearate
: Merck
- CAS No.:112-61-8
- Storage conditions:RT
- Shipping conditions:STD
- MDL No.:MFCD00009005
- M-Clarity quality level:MQ100
- Synonym(s):Methyl octadecanoate; Stearic acid methyl ester
- Melting point:37 - 41 °C (lit.)
- Formula:CH₃(CH₂)₁₆CO₂CH₃
- Molecular weight:298,50
- Flash point:110,0 °C - Closed cup
- Boiling point:181 - 182 °C/4 mmHg (lit.)
Frequently Bought Together