tert-Butyl formate
Catalog # EHERC13913305
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:102.13 g/mol
Storage Temperature:Refrigerator
EINECS:212-105-0
UN:3272
ADR:3,II
:GHS02
:GHS07
CAS Number:762-75-4
Formula:C₅H₁₀O₂