Supelco® Organic Reference Standard, Methyl dodecanoate
61689-5ML
: Merck
:
- Pk:5 mL
- Standards:Analytical standard
- CAS No.:111-82-0
- Description:Methyl dodecanoate
- Storage conditions:RT
- Hazard class description:H410
- Shipping conditions:STD
- MDL No.:MFCD00008966
- M-Clarity quality level:MQ100
- Synonym(s):Lauric acid methyl ester; Methyl laurate
- Melting point:4.5 - 5 °C
- Formula:CH₃(CH₂)₁₀COOCH₃
- Molecular weight:214,34
- Density:0,869 g/ml at 20 °C (lit.)
- Flash point:139 °C - Closed cup
- Boiling point:261 - 262 °C (lit.)
- GHS signal: GHS09
: P273