Order Entry
Denmark
Orders LinkContactUsLinkComponent
Hydroxyurea

Hydroxyurea

Glass bottle for solids, 200 mg
Katalog # PROOBP197
Leverandør:   LGC Standards PROMOCHEM
Hydroxyurea
Katalog # PROOBP197
CAS nummer:  127-07-1
Hydroxyurea

Glass bottle for solids, 200 mg

CAS nummer:  127-07-1
Price
3.740,00 kr
Log ind for tilgængelighed
1 * 200 mg
Product Details & Documents
Synonymer:  N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
  • CAS nummer:127-07-1
  • Formel:CH₄N₂O₂
  • MDL nummer:MFCD00007943
  • Molecular Weight:76.05 g/mol
  • Merck Index:13,04874
  • Kogende Pt:296 °C (1013 hPa)
  • Smeltende Pt:142…144 °C
  • Opbevaringstemperatur:Ambient
  • EINECS:204-821-7
Specifikationer

Specifikationer

  • Pack type:
    Glass bottle for solids
  • Pk:
    200 mg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.