Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Hydroxyurea 98%

Hydroxyurea 98%
 
Hydroxyurea 98%
Hydroxyurea 98%

 

 
 :  N-(Aminocarbonyl)hydroxylamine, Hydroxycarbamide
  •  :127-07-1
  •  :CH₄N₂O₂
  •  :MFCD00007943
  •  :76.05 g/mol
  •  :13,04874
  •  :296 °C (1013 hPa)
  •  :142…146 °C (dec.)
  •  :Refrigerator
  •  :204-821-7

 

 

 

  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    142 to 146 °C
  • Titration Iodometric:
    ≥97.5 % (on dried substance)
  • Loss on drying:
    ≤1 % (1 g, 60 °C) (< 0.7 kPa)
Product Family Options

 

  • Pack type
    Pk
     
     
  • Glass bottle for solids
    5 g
       
       
    1 * 5 g
    1 * 5 g - 842,00 kr
    842,00 kr
    1 * 5 g
       842,00 kr

  • Glass bottle for solids
    25 g
       
       
    1 * 25 g
    1 * 25 g - 3.110,00 kr
    3.110,00 kr
    1 * 25 g
       3.110,00 kr

  • Glass bottle
    100 g
       
       
    1 * 100 g
    1 * 100 g - 7.060,00 kr
    7.060,00 kr
    1 * 100 g
       7.060,00 kr