Order Entry
Denmark
Orders LinkContactUsLinkComponent
Formamide 99.5% for analysis

Formamide 99.5% for analysis

Health
Formamide 99.5% for analysis
Certificates
Formamide 99.5% for analysis

4 Options Available Below

Product Details & Documents
Synonyms:  Carbamaldehyde
  • CAS Number:75-12-7
  • Formula:HCONH₂
  • MDL Number:MFCD00007941
  • Molecular Weight:45.04 g/mol
  • Merck Index:13,04261
  • Boiling Pt:210 °C (760 mmHg)
  • Melting Pt:2…3 °C
  • Density:1,133 g/cm³
  • Flash Pt:175 °C
  • Storage Temperature:Stue temperatur
  • EINECS:200-842-0

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.05 % (Coulometric)
  • Sulfated ash:
    ≤0.005 %
  • GC:
    ≥99.5 %
  • Color scale:
    ≤10 APHA
  • Copper (Cu):
    ≤1 ppm
  • Iron (Fe):
    ≤1 ppm
  • Zinc (Zn):
    ≤1 ppm
  • Heavy metals (as Pb):
    ≤1 ppm
Product Family Options

Product Information

  • Pakning
    Pakn
    Availability
    Price
  • Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 214,00 kr
    214,00 kr
    1 * 100 mL
       214,00 kr

  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 627,00 kr
    627,00 kr
    1 * 1 L
       627,00 kr

  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 1.210,00 kr
    1.210,00 kr
    1 * 2,5 L
       1.210,00 kr

  • Plastic drum
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 3.850,00 kr
    3.850,00 kr
    1 * 10 L
       3.850,00 kr