

Acetone ≥99.5% NF, FCC, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:67-64-1
:
Formula:CH₃COCH₃
MDL Number:MFCD00008765
:58.08 g/mol
Merck Index:13,00067
Boiling Pt:56.2 °C (1013 hPa)
Melting Pt:–95.4 °C
Density:0.792 g/cm³ (20 °C)
Flash Pt:<–20 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
REACH:01-2119471330-49
:GHS02
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Specifications
Specification Test Results
- Bulk Pharmaceutical Chemical:Passes Test
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Benzene:≤ 2 ppm
- NF - Specific Gravity at 25°/25°C:≤ 0.788
- NF - Nonvolatile Residue:≤ 10 ppm
- NF - Readily Oxidizable Substances:Passes Test
- NF - Water (H₂O):≤ 0.5 %
- NF - Assay (as (CH₃)₂CO) (calculated on anhydrous basis):99.5 - 100.5 %
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- FCC - Solubility in H₂O:Passes Test
- FCC - Substances Reducing Permanganate:Passes Test
- FCC - Acidity (as CH₃COOH):≤ 0.002 %
- FCC - Aldehydes (as HCHO):≤ 0.002 %
- FCC - Alkalinity (as NH₃):≤ 10 ppm
- FCC - Lead (Pb):≤ 1 mg/kg
- FCC - Methanol (CH₃OH):≤ 0.05 %
- FCC - Phenols:Passes Test
- FCC - Refractive Index at 20°C:1.358 - 1.360
- FCC - Nonvolatile Residue:≤ 10 ppm
- FCC - Water (H₂O):≤ 0.5 %
- FCC - Distilling Range:≤ 1 °C
Specification Test Results
Related Information
Certificates
Search by Lot NumberBAKR9008-01_TDS_EN BAKR9008-03_TDS_EN spec_BAKR9008-R
Product specification
Frequently Bought Together