Glutaraldehyde
Supplier: Roth Carl
Certificates
About this item
Synonyms: Glutaral, Glutardialdehyde, 1,5-Pentanedial
CAS Number:111-30-8
:
Formula:C₅H₈O₂
MDL Number:MFCD00007025
Boiling Pt:188
Melting Pt:–10 °C
Density:1.014 g/cm³ (20 °C)
Molecular Weight:100.12 g/mol
Storage Temperature:Freezer
EINECS:203-856-5
ADR:6.1 II
Merck Index:13,04485
UN:2927
:GHS06
:GHS05
:GHS08
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together