Propylene oxide ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: (±)-Methyloxirane, (±)-Propene oxide, (±)-Propylene oxide
CAS Number:75-56-9
:
Formula:C₃H₆O
MDL Number:MFCD00005126
Boiling Pt:34°C
Molecular Weight:58.08 g/mol
Melting Pt:-112
Flash Pt:–37°C
ADR:3 I
Merck Index:14,07856
EINECS:200-879-2
UN:1280
:GHS02
:GHS06
:GHS08
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Frequently Bought Together