Order Entry
Denmark
Orders LinkContactUsLinkComponent
Propylene oxide ≥99%

Propylene oxide ≥99%

Flammable
Health
Irritant
Propylene oxide ≥99%
Certificates
Propylene oxide ≥99%
Propylene oxide ≥99%

3 Options Available Below

Product Details & Documents

Purity: 99+% CAS No.: 75-56-9 Molecular Formula: C3H6O Formula Weight: 58.08 Flash Point: -37°(-35°F) Physical Form: Liquid Density: 0.83 Refractive Index: 1.3660 Appearance: Colourless Odour: Ether-like Melting Point: -112° Boiling Point: 34-35° Beilstein Reference: 79763 Merck Reference: 14,7856 UN No.: 1280 MDL No.: MFCD00005126

Synonyms:  (±)-Methyloxirane, (±)-Propene oxide, (±)-Propylene oxide
  • CAS Number:75-56-9
  • Formula:C₃H₆O
  • MDL Number:MFCD00005126
  • Molecular Weight:58.08 g/mol
  • Merck Index:14,7856
  • Boiling Pt:34…35 °C
  • Melting Pt:–112 °C
  • Density:0.83
  • Flash Pt:–37 °C (–35 °F)
  • Storage Temperature:Refrigerator
  • EINECS:200-879-2
  • UN:1280
  • ADR:3,I
Product Family Options

Product Information

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.