Order Entry
Denmark
Orders LinkContactUsLinkComponent
1-Bromo-2-chloroethane

1-Bromo-2-chloroethane

Supplier:   Restek
1-Bromo-2-chloroethane
Supplier:  Restek
Certificates
1-Bromo-2-chloroethane

2 Options Available Below

Product Details & Documents
Synonyms:  Ethylene chlorobromide
  • CAS Number:107-04-0
  • Formula:BrH₂CCH₂Cl
  • MDL Number:MFCD00000962
  • Molecular Weight:143.41 g/mol
  • Boiling Pt:107 °C (1013 hPa)
  • Melting Pt:–17 °C
  • Density:1.7392 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:203-456-0
  • UN:2810
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass ampoule
    1 Ampoul
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each

  • Glass ampoule
    1 Ampoul
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each