Order Entry
Denmark
Orders LinkContactUsLinkComponent
L(+)-Histidine, TECHNICAL

L(+)-Histidine, TECHNICAL

Plastic bottle for solids, 25 g
Catalog # 24581.134
Supplier:   VWR Chemicals
L(+)-Histidine, TECHNICAL
Catalog # 24581.134
Supplier:  VWR Chemicals
CAS Number:  71-00-1
L(+)-Histidine, TECHNICAL
L(+)-Histidine, TECHNICAL

Plastic bottle for solids, 25 g

CAS Number:  71-00-1
Price
426,00 kr
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  L-3-Imidazol-4-ylalanine, (S)-(+)-Histidine, (S)-2-Amino-3-(4-imidazolyl)propionic acid, H-His-OH, NSC 137773, L-Histidine
  • CAS Number:71-00-1
  • Formula:C₆H₉N₃O₂
  • MDL Number:MFCD00064315
  • Molecular Weight:155.16 g/mol
  • Merck Index:13,04741
  • Melting Pt:272…273 °C
  • Density:1.44 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-745-3
Specifications

Specification Test Results

  • Identification:
    Passes test

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.