Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

L(+)-Histidine, Millipore®

L(+)-Histidine, Millipore®
Supplier:  Merck
Certificates
L(+)-Histidine, Millipore®
L(+)-Histidine, Millipore®

3 Options Available Below

Product Details & Documents

Soluble in water and mineral acids; insoluble in organic solvents.

Synonyms:  L-3-Imidazol-4-ylalanine, (S)-(+)-Histidine, (S)-2-Amino-3-(4-imidazolyl)propionic acid, H-His-OH, NSC 137773, L-Histidine
  • CAS Number:71-00-1
  • Formula:C₆H₉N₃O₂
  • MDL Number:MFCD00064315
  • Molecular Weight:155.16 g/mol
  • Merck Index:13,04741
  • Melting Pt:272…273 °C
  • Density:1.44 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ100
  • Storage Temperature:Ambient
  • EINECS:200-745-3
Product Family Options

Product Information