Order Entry
Denmark
ContactUsLinkComponent
L(+)-Histidine
L(+)-Histidine
Catalog # A1341.1000
Supplier:  PanReac AppliChem
CAS Number:  
undefined
L(+)-Histidine
Catalog # A1341.1000
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :155.16 g/mol
Merck Index:13,04741
Melting Pt:272…273 °C
Density:1.44 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-745-3
CAS Number:71-00-1
Formula:C₆H₉N₃O₂
MDL Number:MFCD00064315