D-(-)-Sorbitol 97%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
- CAS Number:50-70-4
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00004708
- Molecular Weight:182.17 g/mol
- Merck Index:13,08797
- Boiling Pt:295 °C (1013 hPa)
- Melting Pt:95…99 °C
- Density:1,524 g/cm³ (20 °C)
- Flash Pt:≥ 100 °C
- Storage Temperature:Ambient
- EINECS:200-061-5
6 Options Available Below
Product Details & Documents
Synonyms: (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
- CAS Number:50-70-4
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00004708
- Molecular Weight:182.17 g/mol
- Merck Index:13,08797
- Boiling Pt:295 °C (1013 hPa)
- Melting Pt:95…99 °C
- Density:1,524 g/cm³ (20 °C)
- Flash Pt:≥ 100 °C
- Storage Temperature:Ambient
- EINECS:200-061-5
Documents
Specifications for Product Family
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Water:≤1.5 %
- Sulfated ash:≤0.1 %
- HPLC:≥96.0 % (on anhydrous substance)
- Specific optical rotation:+4.0° to +7.0° (20°C, 589 nm) (c=10, borate soln.) (on anhydrous substance)
- Lead (Pb):≤0.5 ppm
- Nickel (Ni):≤1 ppm
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



