Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

D-(-)-Mannitol ≥96%, powder

D-(-)-Mannitol ≥96%, powder
Supplier:  MP Biomedicals
Certificates
D-(-)-Mannitol ≥96%, powder
D-(-)-Mannitol ≥96%, powder

3 Options Available Below

Product Details & Documents

Mannitol is carbohydrate found in plants and plant exudates.

A sugar alcohol sweet tastant. Used in sweetness inhibition studies.

A sugar alcohol sweet tastant. Used in sweetness inhibition studies.

Soluble in water (1 g/5,5 ml), alcohol (1 g/83 ml), glycerol (1 g/18 ml); more soluble in hot water; soluble in pyridine, aniline, and aqueous solutions of alkalies; insoluble in ether.

Synonyms:  Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:69-65-8
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064287
  • Molecular Weight:182.17 g/mol
  • Merck Index:13,05769
  • Boiling Pt:290…295 °C (4 hPa)
  • Melting Pt:163…172 °C
  • Density:1.49 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-711-8

 

Product Family Options

Product Information