Order Entry
Denmark
ContactUsLinkComponent
D-(-)-Mannitol 97.0-102.0%, powder, GenAR® USP for biotechnology, Macron Fine Chemicals™
D-(-)-Mannitol 97.0-102.0%, powder, GenAR® USP for biotechnology, Macron Fine Chemicals™
Supplier:  Avantor
Certificates
undefined
D-(-)-Mannitol 97.0-102.0%, powder, GenAR® USP for biotechnology, Macron Fine Chemicals™
Supplier:  Avantor
Certificates

Specification Test Results

  • GMP Manufactured Product:
  • Meets U.S.P Requirements:
  • Meets B.P. Chemical Specifications:
  • Meets E.P. Chemical Specifications:
  • CAUTION: For Manufacturing, processing or repackaging:
  • Bulk Pharmaceutical Chemical:
  • USP - Assay (dried basis):
    97.0 - 102.0 %
  • USP - Identification:
    Passes Test
  • USP - Related Substances: Sorbitol:
    ≤ 2.0 %
  • USP - Related Substances: Sum of isomalt& maltitol:
    ≤ 2.0 %
  • USP - Related Substances: Unspecified impurities:
    ≤ 0.10 %
  • USP - Related Substances: Total impurities:
    ≤ 2.0 %
  • USP - Loss on Drying at 105°C:
    ≤ 0.5 %
  • USP - Melting Point:
    165 - 170 °C
  • USP - Reducing Sugars:
    ≤ 0.1 %
  • USP - Nickel (Ni):
    ≤ 1 ppm
  • USP - Appearance of Solution:
    Passes Test
  • USP - Conductivity (uS cm-1):
    ≤ 20 µS/cm
  • USP - Total Aerobic Microbial Count:
    ≤ 100 cfu/g
  • USP - Total Yeast and Mold Count:
    ≤ 100 cfu/g
  • USP - Escherichia coli:
    Passes Test
  • EP/BP - Assay (as C₆H₁₄O₆) (calculated on anhydrous basis):
    97.0 - 102.0 %
  • EP/BP - Conductivity, uS cm-1:
    ≤ 20
  • EP/BP - Identification C:
    Passes Test
  • EP/BP - Melting Point:
    165 - 170 °C
  • EP/BP - Loss on Drying:
    ≤ 0.5 %
  • EP/BP - Reducing Sugars:
    ≤ 0.1 %
  • EP/BP - Related Substances: Impurity A:
    ≤ 2.0 %
  • EP/BP - Related Substances: Sum of Impurities B & C:
    ≤ 2.0 %
  • EP/BP - Related Substances: Unspecified Impurities, each:
    ≤ 0.10 %
  • EP/BP - Related Substances: Total Impurities:
    ≤ 2.0 %
  • EP/BP - Appearance of Solution:
    Passes Test
  • Total Aerobic Microbial Count (cfu/g):
    ≤ 1000
  • Total Mold and Yeast Count (cfu/g):
    ≤ 100
  • EP/BP - Escherichia coli (absent):
    Passes Test
  • Salmonella (absent):
    Passes Test
  • Endotoxin Concentration (EU/g):
    ≤ 10
  • Appearance (fine, white crystalline powder):
    Passes Test
  • Not Intended for Parenteral Use:
    ≤ 10
  • Only Class 2 solvents (Ethylene Glycol) are likely to be present. All:
  • are below Option 1 limits.:

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together