D-(-)-Mannitol ≥97.0%, white powder
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 69-65-8 Molecular Formula: C6H14O6 Formula Weight: 182.17 Odour: Odourless Melting Point: 165-167° Boiling Point: 290-295°/3.5mm Beilstein Reference: 1721898 Merck Reference: 14,5745 Optical Rotation: +24° (in borax solution) MDL No.: MFCD00064287
Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
- CAS Number:69-65-8
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00064287
- Molecular Weight:182.17 g/mol
- Merck Index:14,5745
- Boiling Pt:290…295 °C (3.5 mmHg)
- Melting Pt:165…167 °C
- Density:1.489
- Storage Temperature:Ambient
- EINECS:200-711-8
3 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 69-65-8 Molecular Formula: C6H14O6 Formula Weight: 182.17 Odour: Odourless Melting Point: 165-167° Boiling Point: 290-295°/3.5mm Beilstein Reference: 1721898 Merck Reference: 14,5745 Optical Rotation: +24° (in borax solution) MDL No.: MFCD00064287
Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
- CAS Number:69-65-8
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00064287
- Molecular Weight:182.17 g/mol
- Merck Index:14,5745
- Boiling Pt:290…295 °C (3.5 mmHg)
- Melting Pt:165…167 °C
- Density:1.489
- Storage Temperature:Ambient
- EINECS:200-711-8



