Order Entry
Denmark
Orders LinkContactUsLinkComponent
D-(-)-Mannitol ACS
D-(-)-Mannitol ACS
D-(-)-Mannitol ACS
D-(-)-Mannitol ACS
CAS No.: 69-65-8 Molecular Formula: HOCH2(CHOH)4CH2OH Formula Weight: 182.17 Physical Form: Crystalline Density: 1.489 Appearance: White Odour: Odourless Melting Point: 165-167° Boiling Point: 290-295°/3.5mm Solubility: Soluble in water, alcohol, pyridine, aniline. Insoluble in ether Beilstein Reference: 1721898 Merck Reference: 14,5745 MDL No.: MFCD00064287
 :  Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
  •  :69-65-8
  •  :HOH₂C(CH(OH))₄CH₂OH
  •  :MFCD00064287
  •  :182.17 g/mol
  •  :14,5745
  •  :290…295 °C (3.5 mmHg)
  •  :165…167 °C
  •  :1.489
  •  :Ambient
  •  :200-711-8
Product Family Options
  • Pack type
    Pk
  • Glass bottle for solids
    500 g
    1 * 500 g
    1 * 500 g - 962,00 kr
    962,00 kr
    1 * 500 g
       962,00 kr
  • Glass bottle for solids
    2,5 kg
    1 * 2,5 kg
    1 * 2,5 kg - 3.820,00 kr
    3.820,00 kr
    1 * 2,5 kg
       3.820,00 kr