Order Entry
Denmark
Orders LinkContactUsLinkComponent
D-(-)-Mannitol ACS

D-(-)-Mannitol ACS

D-(-)-Mannitol ACS
Certifikater
D-(-)-Mannitol ACS

3 tilgængelige muligheder nedenfor

Product Details & Documents
Synonymer:  Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
  • CAS nummer:69-65-8
  • Formel:HOH₂C(CH(OH))₄CH₂OH
  • MDL nummer:MFCD00064287
  • Molecular Weight:182.17 g/mol
  • Merck Index:13,05769
  • Kogende Pt:290 - 295 °C (4 hPa)
  • Smeltende Pt:165…170 °C
  • Tæthed:1,49 g/cm³ (20 °C)
  • Opbevaringstemperatur:Ambient
  • EINECS:200-711-8

Documents

Specifications for Product Family

Specifikationstestresultater

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.05 % (5 g, 105°C, 2 hrs)
  • Residue after ignition:
    ≤0.01 %
  • Specific optical rotation:
    +23.3° to +24.3° (25°C, 589 nm)
  • Heavy metals (ICP-OES):
    ≤5 ppm (as Pb)
  • Titrable acid:
    ≤0.0008 meq/g
  • Insoluble matter:
    ≤0.01 %
  • Reducing sugars:
    Passes test
Product Family Options

Produktinformation

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    250 g
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 250 g
    1 * 250 g - 464,00 kr
    464,00 kr
    1 * 250 g
       464,00 kr

  • Plastic bottle
    1 kg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 kg
    1 * 1 kg - 1.280,00 kr
    1.280,00 kr
    1 * 1 kg
       1.280,00 kr

  • Plastic drum
    5 kg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 5 kg
    1 * 5 kg - 4.280,00 kr
    4.280,00 kr
    1 * 5 kg
       4.280,00 kr

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.