Organic reference standard, 2,2'-Dichlorobiphenyl (PCB No. 4) 10 µg/ml in isooctane, Dr Ehrenstorfer
EHERL20000400IO
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:13029-08-8
- Description:2,2'-Dichlorobiphenyl (PCB No. 4) 10 µg/ml in isooctane
- Formula:C₁₂H₈Cl₂
- Description:2,2'-Dichlorobiphenyl (PCB No. 4)